Azelaic acid ≥96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid
042587-14
042587-22
042587-36
AA42587-14EA
36
USD
AA42587-14
AA42587-22
AA42587-36
Azelaic acid ≥96%
Azelaic acid
MDL: MFCD00004432
Beilstein Registry No.: 1101094
Purity: 96%
CAS No.: 123-99-9
Molecular Formula: HOOC(CH2)7COOH
Formula Weight: 188.22
Flash Point: 215°(419°F)
Physical Form: Crystalline
Density: 1.225
Melting Point: ca 102°
Boiling Point: 286°/100mm
Beilstein Reference: 1101094
Merck Reference: 14,905
MDL No.: MFCD00004432
Formula:
C₉H₁₆O₄ Boiling Pt: 286 °C (100 mmHg) Melting Pt: ∼ 102 °C Density: 1.0287 g/cm³ (20 °C) Flash Pt: 215 °C (419 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004432 CAS Number: 123-99-9 EINECS: 204-669-1 Merck Index: 13,00908 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...