Tellurium ≥99.9999% (metals basis), shot, Diameter 2-5 mm (0.08-0.20 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
042213-06
042213-14
042213-22
AA42213-06EA
64.8
USD
AA42213-06
AA42213-14
AA42213-22
Tellurium ≥99.9999% (metals basis), shot, Diameter 2-5 mm (0.08-0.20 in)
Tellurium
MDL: MFCD00134062
Purity: 99.9999%
CAS No.: 13494-80-9
Odor: Odorless
MDL No.: MFCD00134062
Formula:
Te MW: 127.6 g/mol Boiling Pt: 990 °C (1013 hPa) Melting Pt: 450 °C Density: 8.83 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 EINECS: 236-813-4 UN: 3284 ADR: 6.1,III Merck Index: 12,09273 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...