Palladium silver alloy (75:25 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
042187-FI
042187-FF
042187-GH
AA42187-FIEA
1200
USD
AA42187-FI
AA42187-FF
AA42187-GH
Palladium silver alloy (75:25 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness 0.025 mm (0.001 in)
Palladium silver alloy (75:25 w%)
MDL: MFCD02091738
Purity: 99.9%
Molecular Formula: Pd:Ag; 75:25 wt%
Physical Form: ≈0.18g/25x25mm
Odor: Odorless
MDL No.: MFCD02091738
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...