Iron ≥99.8% (metals basis), sponge -50/+100 mesh

Supplier: Thermo Scientific Chemicals
Warning

041830-22 041830-36
AA41830-22EA 19 USD
AA41830-22 AA41830-36
Iron ≥99.8% (metals basis), sponge -50/+100 mesh
Iron

MDL: MFCD00010999


Purity: 99.8% CAS No.: 7439-89-6 Appearance: Pale to dark grey MDL No.: MFCD00010999

Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR