Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis), foil, Thickness 0.1 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
041804-FF
041804-FI
AA41804-FFEA
374
USD
AA41804-FF
AA41804-FI
Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis), foil, Thickness 0.1 mm (0.004 in)
Platinum iridium alloy (90:10 w%)
MDL: MFCD02091683
Purity: 99.9%
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈1.35g/25x25mm
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...