Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis excluding precious metals), foil, Thickness 0.05 mm (0.002 in)
Supplier: Thermo Scientific Chemicals
041803-FF
041803-FI
AA41803-FFEA
291
USD
AA41803-FF
AA41803-FI
Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis excluding precious metals), foil, Thickness 0.05 mm (0.002 in)
Platinum iridium alloy (90:10 w%)
MDL: MFCD02091683
Purity: 99.9%
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈0.67g/25x25mm
Odor: Odorless
MDL No.: MFCD02091683
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...