Calcium hexaboride ≥99.5%

Supplier: Thermo Scientific Chemicals
Warning

041722-06 041722-14 041722-22
AA41722-06EA 34.9 USD
AA41722-06 AA41722-14 AA41722-22
Calcium hexaboride ≥99.5%
Calcium hexaboride

MDL: MFCD00148904


Purity: 99.5% CAS No.: 12007-99-7 Molecular Formula: CaB6 Formula Weight: 104.95 Physical Form: -325 Mesh Powder Density: 2.30 Melting Point: 2235° MDL No.: MFCD00148904

Formula: B₆Ca
MW: 104.94 g/mol
Melting Pt: 2235 °C
Density: 2.33 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148904
CAS Number: 12007-99-7
EINECS: 234-525-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR