Calcium hexaboride ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
041721-30
041721-18
041721-A1
AA41721-30EA
145
USD
AA41721-30
AA41721-18
AA41721-A1
Calcium hexaboride ≥95%
Calcium hexaboride
MDL: MFCD00148904
Purity: 95+%
CAS No.: 12007-99-7
Molecular Formula: CaB6
Formula Weight: 104.95
Physical Form: -20 Mesh Powder
Density: 2.30
Melting Point: 2235°
MDL No.: MFCD00148904
Formula:
B₆Ca MW: 104.94 g/mol Melting Pt: 2000 °C Density: 2.33 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00148904 CAS Number: 12007-99-7 EINECS: 234-525-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...