Silver ≥99.9% (metals basis), spherical powder APS 0.5-1 µm
Supplier: Thermo Scientific Chemicals
041597-18
041597-30
041597-09
AA41597-18EA
282
USD
AA41597-18
AA41597-30
AA41597-09
Silver ≥99.9% (metals basis), spherical powder APS 0.5-1 µm
Silver
MDL: MFCD00003397
Purity: 99.9%
CAS No.: 7440-22-4
Physical Form: S.A. 1.5-2.5m2/g
Odor: Characteristic
UN No.: 3077
MDL No.: MFCD00003397
Formula:
Ag Boiling Pt: 2212 °C (1013 hPa) Melting Pt: 961.9 °C Density: 10.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...