Digallium trisulfide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gallium (III) sulphide

041538-06 041538-SS
AA41538-06EA 325 USD
AA41538-06 AA41538-SS
Digallium trisulfide ≥99.999% (metals basis)
Digallium trisulphide

MDL: MFCD00016106


Purity: 99.999% CAS No.: 12024-22-5 Molecular Formula: Ga2S3 Formula Weight: 235.63 Physical Form: Powder Density: 3.77 Melting Point: 1255° UN No.: 3260 MDL No.: MFCD00016106

Formula: Ga₂S₃
MW: 235.64 g/mol
Melting Pt: 1255 °C
Density: 3.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016106
CAS Number: 12024-22-5
EINECS: 234-692-2
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR