Yttrium(III) fluoride, anhydrous ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

041424-09 041424-30 041424-18
AA41424-09EA 72.6 USD
AA41424-09 AA41424-30 AA41424-18
Yttrium(III) fluoride, anhydrous ≥99.9% (metals basis)
Yttrium(III) fluoride

For vacuum deposition


Purity: 99.9% CAS No.: 13709-49-4 Molecular Formula: YF3 Formula Weight: 145.90 Physical Form: 3-12mm Pieces Sensitivity: Hygroscopic UN No.: 3288 MDL No.: MFCD00049714


For vacuum deposition

Formula: F₃Y
Boiling Pt: 2227 °C (1013 hPa)
Melting Pt: 1387 °C
Density: 4.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049714
CAS Number: 13709-49-4
EINECS: 237-257-5
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR