Aluminium ≥98% (metals basis), spherical powder APS 10.0-14.0 µm
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminum
041001-30
041001-A7
041001-A1
AA41001-30EA
56.72
USD
AA41001-30
AA41001-A7
AA41001-A1
Aluminium ≥98% (metals basis), spherical powder APS 10.0-14.0 µm
Aluminium
MDL: MFCD00134029
Notes: Tap density 1.5g/cm3
Purity: 98%
CAS No.: 7429-90-5
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Grey
Odor: Odorless
UN No.: 1396
MDL No.: MFCD00134029
Formula:
Al MW: 26.98 g/mol Boiling Pt: 2057 °C (1013 hPa) Melting Pt: 659.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 UN: 1396 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...