Aluminium ≥99.995% (metals basis), ultrathin foil, Thickness: 0.0075 micron
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminum
040682-DH
040682-HF
AA40682-DHEA
915
USD
AA40682-DH
AA40682-HF
Aluminium ≥99.995% (metals basis), ultrathin foil, Thickness: 0.0075 micron
Aluminium
Microelectronics, contacts and coatings
Purity: 99.995%
CAS No.: 7429-90-5
Physical Form: Circular disc on permanent mylar support
Odor: Odorless
MDL No.: MFCD00134029
Microelectronics, contacts and coatings
Formula:
Al MW: 26.98 g/mol Boiling Pt: 2327 Melting Pt: 660 Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...