Tantalum ≥99.9% (metals basis), wire, hard, Diameter 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals

040675-G6 040675-G1
AA40675-G5EA 402.26 USD
AA40675-G5 AA40675-G6 AA40675-G1
Tantalum ≥99.9% (metals basis), wire, hard, Diameter 0.025 mm (0.001 in)
Tantalum

Microelectronics, contacts, surgical, high temperature


Purity: 99.9% CAS No.: 7440-25-7 Physical Form: ≈0.0082g/m, Temper: as drawn Odor: Odorless MDL No.: MFCD00011252


Microelectronics, contacts, surgical, high temperature

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR