Indium ≥99.998% (metals basis), wire, Puratronic®, Diameter 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

040421-G9 040421-G3 040421-G6
AA40421-G9EA 583 USD
AA40421-G9 AA40421-G3 AA40421-G6
Indium ≥99.998% (metals basis), wire, Puratronic®, Diameter 0.25 mm (0.01 in)
Indium

As solders, thermal fuses, fusible alloys, and in semiconductor industry in InAs and InSb forms


Purity: 99.998+% CAS No.: 7440-74-6 Physical Form: ≈0.36g/m Odor: Odorless MDL No.: MFCD00134048


As solders, thermal fuses, fusible alloys, and in semiconductor industry in InAs and InSb forms

Formula: In
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR