Sodium iodate ≥99%

Supplier: Thermo Scientific Chemicals
Danger

040135-22 040135-36
AA40135-22EA 70 USD
AA40135-22 AA40135-36
Sodium iodate ≥99%
Sodium iodate

MDL: MFCD00003533


Purity: 99% min CAS No.: 7681-55-2 Molecular Formula: NaIO3 Formula Weight: 197.89 Physical Form: Crystalline powder Density: 4.28 Appearance: White Melting Point: dec. Merck Reference: 14,8630 UN No.: 1479 MDL No.: MFCD00003533

Formula: INaO₃
MW: 197.89 g/mol
Melting Pt: Dec.
Density: 3.56 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003533
CAS Number: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1,II
Merck Index: 13,08704
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=16d5efbf45a7cc1cd6399272211e2455e47a2e4b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR