Sodium metabisulphite ≥97% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Disodium disulphite, di-Sodium disulphite, Sodium pyrosulphite, Disodiumdisulphite, Sodium disulphite

040115-A4 040115-36
AA40115-A4EA 143 USD
AA40115-A4 AA40115-36
Sodium metabisulphite ≥97% ACS
Sodium metabisulphite

MDL: MFCD00167602


Purity: 97% min CAS No.: 7681-57-4 Molecular Formula: Na2O5S2 Formula Weight: 190.10 Physical Form: Crystalline powder Density: 1.480 Appearance: White to pale yellow Odor: Like rotten eggs pH: 3.5-5.0 Melting Point: 300° Merck Reference: 14,8638 MDL No.: MFCD00167602

Formula: Na₂S₂O₅
MW: 190.11 g/mol
Melting Pt: 150
Density: 1.480
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
REACH: 01-2119531326-45
Merck Index: 14,8638
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.005%
Cl 0.05%
Heavy Metals (as Pb) 0.001%
Fe 0.002%
S2O3 0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR