Fullerene C70 97%, powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
039720-MA
039720-MF
039720-MD
AA39720-MAEA
62.3
USD
AA39720-MA
AA39720-MF
AAA39720-MD
Fullerene C70 97%, powder
Fullerene C70
MDL: MFCD00282904
Purity: 97%
CAS No.: 115383-22-7
Density: 1.7
Appearance: Black
Boiling Point: 500-600° subl.
UN No.: 1325
MDL No.: MFCD00282904
Formula:
C₇₀ Boiling Pt: 500…600 °C (subl.) Melting Pt: ? 280 °C Density: 1.73 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00282904 CAS Number: 115383-22-7 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...