Tantalum ≥99.9% (metals basis), powder APS ∼2 µm

Supplier: Thermo Scientific Chemicals
Warning

039690-06 039690-14 039690-18
AA39690-06EA 87.55 USD
AA39690-06 AA39690-14 AA39690-18
Tantalum ≥99.9% (metals basis), powder APS ∼2 µm
Tantalum

MDL: MFCD00011252


Purity: 99.9% CAS No.: 7440-25-7 Appearance: Black Odor: Odorless UN No.: 3089 MDL No.: MFCD00011252

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
UN: 3089
ADR: 4.1 II
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR