Copper ≥99.996% (metals basis), slug, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

039675-14 039675-22
AA39675-14EA 46.5 USD
AA39675-14 AA39675-22
Copper ≥99.996% (metals basis), slug, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Copper

MDL: MFCD00010965


Purity: 99.996% CAS No.: 7440-50-8 Physical Form: ≈1.8g/slug see TECH Notes Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR