Sodium stearate

Supplier: Thermo Scientific Chemicals

Synonyms: Stearate sodium, Stearic acid sodium salt

039648-A1 039648-A7 039648-30
AA39648-A1EA 103.47 USD
AA39648-A1 AA39648-A7 AAA39648-30
Sodium stearate
Sodium stearate

MDL: MFCD00036404 Slowly soluble in cold water. Solubility increases with temperature


CAS No.: 822-16-2 Molecular Formula: NaCO2(CH2)16CH3 Formula Weight: 306.46 Physical Form: Powder Odor: Weak Solubility: Slowly soluble in cold water. Solubility increases with temperature MDL No.: MFCD00036404

Formula: C₁₈H₃₅NaO₂
MW: 306.46 g/mol
Melting Pt: 288
Density: 1.103 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036404
CAS Number: 822-16-2
EINECS: 212-490-5
Merck Index: 14,8678

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR