Tin ≥99.5%, mossy ACS

Supplier: Thermo Scientific Chemicals

039640-30 039640-18
AA39640-30EA 76.69 USD
AA39640-30 AA39640-18
Tin ≥99.5%, mossy ACS
Tin

MDL: MFCD00133862


Purity: 99.5% CAS No.: 7440-31-5 Density: 7.310G/CC Odor: Odorless Melting Point: 231.9 deg C Boiling Point: 2270 deg C MDL No.: MFCD00133862

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now

Specification Test Results

Sb 0.02% Max.
As 1ppm Max.
Cu 0.005% Max.
Fe 0.01% Max.
PB 0.005% Max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR