Potassium metavanadate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

039501-22 039501-14
AA39501-22EA 212 USD
AA39501-22 AA39501-14
Potassium metavanadate ≥99.9% (metals basis)
Potassium metavanadate

MDL: MFCD00050038


Purity: 99.9% CAS No.: 13769-43-2 Molecular Formula: KVO3 Formula Weight: 138.04 Physical Form: -200 Mesh Powder Odor: Odorless UN No.: 2864 MDL No.: MFCD00050038

Formula: KO₃V
Melting Pt: 520 °C
Density: 2,84 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00050038
CAS Number: 13769-43-2
EINECS: 237-388-8
UN: 2864
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR