Cesium orthovanadate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

039211-09 039211-18
AA39211-09EA 119 USD
AA39211-09 AA39211-18
Cesium orthovanadate ≥99.9% (metals basis)
Cesium orthovanadate

MDL: MFCD00210609


Purity: 99.9% CAS No.: 34283-69-7 Molecular Formula: Cs3VO4 Formula Weight: 513.65 Physical Form: -100 Mesh Powder UN No.: 3288 MDL No.: MFCD00210609

Formula: Cs₃O₄V
MW: 513.66
Storage Temperature: Ambient
MDL Number: MFCD00210609
CAS Number: 34283-69-7
EINECS: 251-916-4
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR