Thymolphthalein 0,05% (w/v) in ethanol

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one

038706-AE 038706-AP
AA38706-AEEA 37.8 USD
AA38706-AE AA38706-AP
Thymolphthalein 0,05% (w/v) in ethanol
Thymolphthalein

MDL: MFCD00005909 Beilstein Registry No.: 359413 Notes: Transition interval: pH 8.8 (colorless) to pH 10.5 (blue)


CAS No.: 125-20-2 Molecular Formula: C28H30O4 Formula Weight: 430.55 Flash Point: 11°(52°F) Physical Form: Liquid Density: 0.920 Appearance: Colorless Odor: Alcohol-like Beilstein Reference: 359413 Merck Reference: 14,9401 UN No.: 1170 MDL No.: MFCD00005909

Formula: C₂₈H₃₀O₄
Density: 0.920
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005909
CAS Number: 125-20-2
EINECS: 204-729-7
UN: 1170
ADR: 3,II
Merck Index: 13,09478

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR