Methyl red 0.1% (w/v) in ethanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
038698-AE
038698-AP
AA38698-AEEA
31.54
USD
AA38698-AE
AA38698-AP
Methyl red 0.1% (w/v) in ethanol
Methyl red
MDL: MFCD00002425
Notes: Transition interval: pH 4.2 (pink) to pH 6.2 (yellow)
Molecular Formula: (CH3)2NC6H4N=NC6H4CO2H
Formula Weight: 269.30
Physical Form: Liquid
UN No.: 1170
MDL No.: MFCD00002425
Formula:
C₁₅H₁₅N₃O₂ MW: 269.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002425 CAS Number: 493-52-7 EINECS: 207-776-1 UN: 1170 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...