Potassium chlorate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
As an oxidizing agent, in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes
Purity: 99.0% min
CAS No.: 3811-04-9
Molecular Formula: KClO3
Formula Weight: 122.55
Physical Form: Crystalline
Density: 2.32
Appearance: White
Odor: Odorless
Melting Point: 368°
Boiling Point: 400° dec.
Solubility: Soluble in water, glycerol. Insoluble in alcohol
Merck Reference: 14,7620
UN No.: 1485
MDL No.: MFCD00011361
As an oxidizing agent, in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes
Formula:
KClO₃ Boiling Pt: 400 °C (dec.) Melting Pt: 368 °C Density: 2.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011361 CAS Number: 3811-04-9 EINECS: 223-289-7 UN: 1485 ADR: 5.1,II Merck Index: 13,07703 |
Specification Test Results
Insoluble matter | 0.005% |
BrO3 | 0.015% |
Cl | 0.001% |
SO3 | Pass test (limit about 0.002%) |
Ca | 0.002% |
MG | 0.002% |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 3 ppm |
NA | 0.01%% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...