Sodium tetraphenylborate ≥99.5% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt

036430-04 036430-09 036430-18
AA36430-04EA 28.46 USD
AA36430-04 AA36430-09 AA36430-18
Sodium tetraphenylborate ≥99.5% ACS
Sodium tetraphenylborate

MDL: MFCD00011494 Beilstein Registry No.: 3599783


Purity: 99.5% min CAS No.: 143-66-8 Molecular Formula: C24H20BNa Formula Weight: 342.23 Physical Form: Powder Sensitivity: Light Sensitive Appearance: White pH: 8 Melting Point: 300° Beilstein Reference: 3599783 Merck Reference: 14,8690 UN No.: 2811 MDL No.: MFCD00011494

Formula: (C₆H₅)₄BNa
MW: 342.22 g/mol
Melting Pt: >300 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011494
CAS Number: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,08765

Order Now

Specification Test Results

Loss on drying at 150°C 0.5%
Clarity of solution Pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR