Tannic acid ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Gallotannin, Tannin

036410-36 036410-22
AA36410-36EA 202.05 USD
AA36410-36 AA36410-22
Tannic acid ACS
Tannic acid

MDL: MFCD00066397 Beilstein Registry No.: 8186396 Soluble in water warm glycerol alcohol acetone


CAS No.: 1401-55-4 Molecular Formula: C76H52O46 Formula Weight: 1701.23 Flash Point: 199°(390°F) Physical Form: Powder Sensitivity: Air and Light Sensitive pH: 3.5 Melting Point: ca 218° dec. Solubility: Soluble in water, warm glycerol, alcohol, acetone Beilstein Reference: 8186396 Merck Reference: 14,9052 MDL No.: MFCD00066397

Formula: C76H52O46
MW: 1701.2 g/mol
Melting Pt: ∼ 218 °C (dec.)
Density: Min. 1 g/cm³ (20 °C)
Flash Pt: 199 °C (390 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066397
CAS Number: 1401-55-4
EINECS: 215-753-2
Merck Index: 12,09221

Order Now

Specification Test Results

Identification Pass test
Loss on drying 0.12
Residue after ignition 0.5%
Heavy Metals (as Pb) 0.003%
Zn 0.005%
Sugars & dextrin Pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR