Chlorobenzene ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00000530
Beilstein Registry No.: 605632
Purity: 99.5%
CAS No.: 108-90-7
Molecular Formula: C6H5Cl
Formula Weight: 112.56
Flash Point: 28°(82°F)
Physical Form: Liquid
Density: 1.106
Refractive Index: 1.5240
Appearance: Colorless
Odor: Aromatic
Melting Point: -45°
Boiling Point: 132°
Beilstein Reference: 605632
Merck Reference: 14,2121
UN No.: 1134
MDL No.: MFCD00000530
Formula:
C₆H₅Cl Boiling Pt: 132 °C Melting Pt: –45 °C Density: 1.106 Flash Pt: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000530 CAS Number: 108-90-7 EINECS: 203-628-5 UN: 1134 ADR: 3,III REACH: 01-2119432722-45 Merck Index: 14,2121 |
|
Specification Test Results
Color (APHA) | 30 |
Evaporation residue | 0.02% |
Titratable acid | 0.004meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...