Silicon ≥99.9999% (metals basis), polycrystalline rods, unpolished, Diameter 1.6cm (0.6 in)

Supplier: Thermo Scientific Chemicals
Warning

036376-HF 036376-DM
AA36376-HFEA 113 USD
AA36376-HF AA36376-DM
Silicon ≥99.9999% (metals basis), polycrystalline rods, unpolished, Diameter 1.6cm (0.6 in)
Silicon

MDL: MFCD00085311


Purity: 99.9999% CAS No.: 7440-21-3 Physical Form: ≈4.68g/cm Odor: Odorless MDL No.: MFCD00085311

Formula: Si
Boiling Pt: 2355 °C (1013 hPa)
Melting Pt: 1410 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR