Tantalum(V) oxide ≥99.95% (metals basis excluding Nb) (max. 200 ppm Nb)

Supplier: Thermo Scientific Chemicals

036343-14 036343-22
AA36343-14EA 227 USD
AA36343-14 AA36343-22
Tantalum(V) oxide ≥99.95% (metals basis excluding Nb) (max. 200 ppm Nb)
Tantalum(V) oxide

MDL: MFCD00011254 Notes: For vacuum deposition


Purity: 99.95% CAS No.: 1314-61-0 Molecular Formula: Ta2O5 Formula Weight: 441.89 Physical Form: 3-12mm Sintered Lump Density: 8.2 Melting Point: 1800° Merck Reference: 14,9056 MDL No.: MFCD00011254

Formula: O₅Ta₂
Melting Pt: 1800 °C
Density: 8.2
Storage Temperature: Ambient
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2
Merck Index: 14,9056

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR