Mercury(II) sulfate ≥98.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Applications: Primary battery electrolyte, catalyst in conversion of acetylene to acetaldehyde
Purity: 98.0% min
CAS No.: 7783-35-9
Molecular Formula: HgO4S
Formula Weight: 296.65
Physical Form: Powder
Density: 6.470
Appearance: White to pale yellow
Melting Point: dec.
Solubility: Decomposes in water. Soluble in HCl
Merck Reference: 14,5887
UN No.: 1645
MDL No.: MFCD00011047
Primary battery electrolyte, catalyst in conversion of acetylene to acetaldehyde
Formula:
HgSO₄ Melting Pt: Dec. Density: 6.470 Storage Temperature: Ambient |
MDL Number:
MFCD00011047 CAS Number: 7783-35-9 EINECS: 231-992-5 UN: 1645 ADR: 6.1,II Merck Index: 14,5887 |
Specification Test Results
Residue after reduction | 0.02% |
Cl | 0.003% |
NO7 | Pass test |
Fe | 0.005% |
Mercurous mercury (as Hg) | 0.15% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...