Yttrium(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
035746-04
035746-09
AA35746-04EA
140
USD
AA35746-04
AA35746-09
Yttrium(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Yttrium(III) bromide
MDL: MFCD00049710
Purity: 99.9%
CAS No.: 13469-98-2
Molecular Formula: YBr3
Formula Weight: 328.63
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
MDL No.: MFCD00049710
Formula:
Br₃Y Melting Pt: 904 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049710 CAS Number: 13469-98-2 EINECS: 236-728-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...