Zinc iodide, ultra dry ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zinc (II) iodide
035727-06
035727-14
035727-22
AA35727-06EA
85.78
USD
AA35727-06
AA35727-14
AA35727-22
Zinc iodide, ultra dry ≥99.995% (metals basis)
Zinc iodide
MDL: MFCD00011299
Purity: 99.995%
CAS No.: 10139-47-6
Molecular Formula: ZnI2
Formula Weight: 319.18
Physical Form: Granular, Ampouled under argon
Density: 4.74
Melting Point: 446°
Boiling Point: 625° dec.
Merck Reference: 14,10140
UN No.: 3260
MDL No.: MFCD00011299
Formula:
I₂Zn MW: 319.2 g/mol Boiling Pt: 620 (dec.) Melting Pt: 446 Density: 4.74 Storage Temperature: Ambient |
MDL Number:
MFCD00011299 CAS Number: 10139-47-6 EINECS: 233-396-0 UN: 3260 ADR: 8 II Merck Index: 14,10140 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...