Zinc bromide, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc (II) bromide

035725-06 035725-22
AA35725-06EA 80 USD
AA35725-06 AAA35725-22
Zinc bromide, ultra dry ≥99.999% (metals basis)
Zinc bromide

MDL: MFCD00011294 Fieser: 2,463 13,349 15,368 16,389 18,409 19,409 20,438 21,493


Purity: 99.999% CAS No.: 7699-45-8 Molecular Formula: ZnBr2 Formula Weight: 225.19 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 4.201 Refractive Index: 1.5452 Sensitivity: Hygroscopic Melting Point: 394° Boiling Point: 650° Merck Reference: 14,10129 UN No.: 3260 MDL No.: MFCD00011294

Formula: Br₂Zn
MW: 225.2 g/mol
Boiling Pt: 650 °C
Melting Pt: 394
Density: 4.201
Storage Temperature: Ambient
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8 II
Merck Index: 14,10129

Order Now

Specification Test Results

Form -10 Mesh Beads, Ampouled under argon


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR