Indium(I) iodide, ultra dry ≥99.998% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

035700-06 035700-03
AA35700-06EA 306 USD
AA35700-06 AA35700-03
Indium(I) iodide, ultra dry ≥99.998% (metals basis)
Indium(I) iodide

MDL: MFCD00135543


Purity: 99.998% CAS No.: 13966-94-4 Molecular Formula: InI Formula Weight: 241.72 Physical Form: -10 Mesh Beads, Ampouled under argon Sensitivity: Moisture and Light Sensitive MDL No.: MFCD00135543

Formula: InI
MW: 241.72 g/mol
Boiling Pt: 711 °C (1013 hPa)
Melting Pt: 351 °C
Density: 5.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135543
CAS Number: 13966-94-4
EINECS: 237-746-3
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR