Gallium triiodide, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
035699-03
035699-06
035699-14
AA35699-03EA
66.4
USD
AA35699-03
AA35699-06
AA35699-14
Gallium triiodide, ultra dry ≥99.999% (metals basis)
Gallium triiodide
MDL: MFCD00016107
Purity: 99.999%
CAS No.: 13450-91-4
Molecular Formula: GaI3
Formula Weight: 450.43
Physical Form: Powder, Ampouled under argon
Density: 4.15
Appearance: Yellow-green
Melting Point: 212°
Boiling Point: 345° subl.
UN No.: 3260
MDL No.: MFCD00016107
Formula:
GaI₃ MW: 450.44 Boiling Pt: 345 (subl.) Melting Pt: 212 Density: 4.15 Storage Temperature: Ambient |
MDL Number:
MFCD00016107 CAS Number: 13450-91-4 EINECS: 236-611-6 UN: 3260 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...