Dysprosium trichloride, ultra dry ≥99.98% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

035692-14 035692-06
AA35692-14EA 1198 USD
AA35692-14 AA35692-06
Dysprosium trichloride, ultra dry ≥99.98% (REO, rare earth oxide basis)
Dysprosium trichloride

MDL: MFCD00010984


Purity: 99.98% CAS No.: 10025-74-8 Molecular Formula: DyCl3 Formula Weight: 268.86 Physical Form: Powder, Ampouled under argon Density: 3.67 Melting Point: 718° Merck Reference: 14,3482 MDL No.: MFCD00010984

Formula: Cl₃Dy
MW: 268.86 g/mol
Melting Pt: 642 - 652 °C
Density: 3.67 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010984
CAS Number: 10025-74-8
EINECS: 233-039-9
Merck Index: 14,3482

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR