Sulfuric acid standardised solution 0.1 N in aqueous solution
Supplier: Thermo Scientific Chemicals
035651-K7
035651-M4
035651-K2
AA35651-K7EA
158.39
USD
AA35651-K7
AA35651-M4
AA35651-K2
Sulfuric acid standardised solution 0.1 N in aqueous solution
Sulfuric acid
MDL: MFCD00064589
Molecular Formula: H2SO4
Formula Weight: 98.08
Physical Form: Liquid
MDL No.: MFCD00064589
Formula:
H₂SO₄ Density: 1.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 2796 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...