Sulfuric acid standardised solution 0.02 N in aqueous solution

Supplier: Thermo Scientific Chemicals

035649-K2 035649-M4 035649-K7
AA35649-K2EA 75.87 USD
AA35649-K2 AA35649-M4 AA35649-K7
Sulfuric acid standardised solution 0.02 N in aqueous solution
Sulfuric acid

MDL: MFCD00064589


Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid MDL No.: MFCD00064589

Formula: H₂SO₄
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR