Nitric acid solution_standardised 0.1 N
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Nitric acid solution
035626-K2
035626-AP
AA35626-K2EA
50.53
USD
AA35626-K2
AA35626-AP
Nitric acid solution_standardised 0.1 N
Nitric acid
MDL: MFCD00011349
CAS No.: 7697-37-2
Molecular Formula: HNO3
Formula Weight: 63.01
Physical Form: Liquid
Appearance: Colorless
Formula:
HNO₃ MW: 63.01 Boiling Pt: 87 Melting Pt: -42 Density: 1.51 Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 EINECS: 231-714-2 UN: 3264 ADR: 8 III Merck Index: 13,06610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...