Chlorotrimethylgermane
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00000462
CAS No.: 1529-47-1
Molecular Formula: (CH3)3GeCl
Formula Weight: 153.15
Flash Point: 1°(35°F)
Physical Form: Liquid
Density: 1.249
Refractive Index: 1.4337
Sensitivity: Moisture Sensitive
Melting Point: -14°
Boiling Point: 102°
UN No.: 2924
MDL No.: MFCD00000462
Formula:
C₃H₉ClGe Boiling Pt: 102 °C Melting Pt: –14 °C Density: 1.2435 g/cm³ (20 °C) Flash Pt: 1 °C (35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000462 CAS Number: 1529-47-1 EINECS: 216-214-4 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...