Dichlorodimethylgermane 98+%

Supplier: Thermo Scientific Chemicals
Danger

034107-04 034107-09
AA34107-04EA 107.51 USD
AA34107-04 AA34107-09 AA34107-02
Dichlorodimethylgermane 98+%
Dichlorodimethylgermane

MDL: MFCD00013588


Purity: 98+% CAS No.: 1529-48-2 Molecular Formula: (CH3)2GeCl2 Formula Weight: 173.57 Flash Point: 32°(90°F) Physical Form: Liquid Density: 1.51 Refractive Index: 1.460 Sensitivity: Moisture Sensitive Melting Point: -22° Boiling Point: 124° UN No.: 2924 MDL No.: MFCD00013588

Formula: C₂H₆Cl₂Ge
Boiling Pt: 124 °C
Melting Pt: –22 °C
Density: 1.4926 g/cm³ (20 °C)
Flash Pt: 32 °C (90 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013588
CAS Number: 1529-48-2
EINECS: 216-216-5
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR