Triisopropylphosphine 90+%

Supplier: Thermo Scientific Chemicals
Danger

033572-02 033572-09
AA33572-02EA 39.7 USD
AA33572-02 AA33572-09
Triisopropylphosphine 90+%
Triisopropylphosphine

MDL: MFCD00015029


Purity: 90+% CAS No.: 6476-36-4 Molecular Formula: [(CH3)2CH]3P Formula Weight: 160.24 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.839 Refractive Index: 1.4670 Sensitivity: Air Sensitive Odor: Pungent Boiling Point: 176-178° UN No.: 2845 MDL No.: MFCD00015029

Formula: C₉H₂₁P
Boiling Pt: 176…178 °C
Density: 0.839
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015029
CAS Number: 6476-36-4
EINECS: 229-333-1
UN: 2845
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR