Potassium cyanate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cyanic acid potassium salt
033356-22
033356-36
033356-A3
AA33356-22EA
39.85
USD
AA33356-22
AA33356-36
AA33356-A3
Potassium cyanate 97%
Potassium cyanate
MDL: MFCD00011411
Soluble in water. Slightly soluble in alcohol
Purity: 97%
CAS No.: 590-28-3
Molecular Formula: KOCN
Formula Weight: 81.12
Physical Form: Crystalline
Density: 2.056
Appearance: White
Odor: Odorless
Melting Point: 700-900° dec.
Solubility: Soluble in water. Slightly soluble in alcohol
Merck Reference: 14,7625
MDL No.: MFCD00011411
Formula:
CKNO MW: 81.12 g/mol Melting Pt: 700…900 °C (dec.) Density: 2.056 Storage Temperature: Ambient |
MDL Number:
MFCD00011411 CAS Number: 590-28-3 EINECS: 209-676-3 Merck Index: 14,7625 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...