Naphthalene ≥99.6%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Formerly important in dyestuff manufacture. As a precursor to phthalic anhydride
Purity: 99.6%
CAS No.: 91-20-3
Molecular Formula: C10H8
Formula Weight: 128.17
Flash Point: 78°(172°F)
Physical Form: Crystalline
Density: 0.9628
Refractive Index: 1.5821
Odor: Aromatic
Melting Point: 80-82°
Boiling Point: 218°
Solubility: Soluble in benzene, toluene, chloroform, CS2, ether. Insoluble in water
Beilstein Reference: 1421310
Merck Reference: 14,6370
UN No.: 1334
MDL No.: MFCD00001742
Formerly important in dyestuff manufacture. As a precursor to phthalic anhydride
Formula:
C₁₀H₈ MW: 128.17 g/mol Boiling Pt: 218 °C Melting Pt: 80…82 °C Density: 1.145 g/cm³ (20 °C) Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001742 CAS Number: 91-20-3 EINECS: 202-049-5 UN: 1334 ADR: 4.1,III Merck Index: 12,06457 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...