Nitric acid 68,0 - 70,0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nitric acid solution

033260-P2 033260-P7 033260-R1
AA33260-P2EA 77.12 USD
AA33260-P2 AA33260-P7 AA-33260-R1
Nitric acid 68,0 - 70,0% ACS
Nitric acid

MDL: MFCD00011349


CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Density: 1.41 Appearance: Colorless to pale yellow Odor: Pungent Boiling Point: 121° Merck Reference: 14,6577 UN No.: 2031 MDL No.: MFCD00011349

Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.41
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 14,6577

Order Now

Specification Test Results

Color (APHA) 10
Residue after ignition 5 ppm
Cl 0.5 ppm
SO3 1 ppm
As 0.01 ppm
Heavy Metals (as Pb) 0.2 ppm
Fe 0.2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR