Acetaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethanal
033244-AE
033244-AP
033244-AY
AA33244-AEEA
74.94
USD
AA33244-AE
AA33244-AP
AA33244-AY
Acetaldehyde 99%
Acetaldehyde
MDL: MFCD00006991
Beilstein Registry No.: 505984
Purity: 98.5%
CAS No.: 75-07-0
Molecular Formula: CH3CHO
Formula Weight: 44.06
Flash Point: -27°(-17°F)
Physical Form: Liquid
Density: 0.780
Refractive Index: 1.3316
Sensitivity: Air Sensitive
Appearance: Colorless
Odor: Pungent
Melting Point: -128°
Boiling Point: 20.8°
Beilstein Reference: 505984
Merck Reference: 14,39
UN No.: 1089
MDL No.: MFCD00006991
Formula:
CH₃CHO MW: 44.05 g/mol Boiling Pt: 20.1 °C (1013 hPa) Melting Pt: -123 °C Density: 0.78 g/cm³ (20 °C) Flash Pt: -27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006991 CAS Number: 75-07-0 EINECS: 200-836-8 UN: 1089 ADR: 3,I Merck Index: 12,00037 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...