Chloroform ≥99.8% stabilized ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00000826
Beilstein Registry No.: 1731042
Fieser: 1,29 3,140 4,22 5,28 8,92 12,517 15,84
Purity: 99.8+%
CAS No.: 67-66-3
Molecular Formula: CHCl3
Formula Weight: 119.38
Physical Form: Liquid
Density: 1.492
Refractive Index: 1.4460
Odor: Characteristic
Melting Point: -63°
Boiling Point: 60.5-61.5°
Beilstein Reference: 1731042
Merck Reference: 14,2141
UN No.: 1888
MDL No.: MFCD00000826
Formula:
CHCl₃ Boiling Pt: 61.7 °C (1013 hPa) Melting Pt: -63 °C Density: 1.4832 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000826 CAS Number: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1,III REACH: 05-2114614906-45 Merck Index: 13,02160 |
|
Specification Test Results
Color (APHA) | 10 |
Residue after evaporation | 0.001% |
Acetone and aldehyde | Pass test (limit about 0.005% as (CH3)2CO) |
Acid and chlorine | Pass test |
Free chlorine (Cl) | Pass test |
PB | 0.05 ppm |
Substances darkened by sulfuric acid | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...